2-Mercapto-2-thiazolin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol
L04924.22
L04924.36
L04924.22EA
41.9
EUR
L04924.22
L04924.36
2-Mercapto-2-thiazolin ≥98%
2-Mercapto-2-thiazolin
Purity: 98+%
CAS No.: 96-53-7
Molecular Formula: C3H5NS2
Formula Weight: 119.21
Melting Point: 103-107°
Beilstein Reference: 106332
UN No.: 2811
MDL No.: MFCD00126013
Formel:
C₃H₅NS₂ Schmelzpunkt: 103…107 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00126013 CAS-Nummer: 96-53-7 EINECS: 202-512-1 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...