Diethylphenylphosphonit ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B24836.06 B24836.14 B24836.22
B24836.06EA 140 EUR
B24836.06 B24836.14 B24836.22
Diethylphenylphosphonit ≥98%
Diethylphenylphosphonit

Purity: 98% CAS No.: 1638-86-4 Molecular Formula: (CH3CH2O)2P(C6H5) Formula Weight: 198.20 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5100 Boiling Point: 64°/0.7mm Beilstein Reference: 2804900 MDL No.: MFCD00009086

Formel: C₁₀H₁₅O₂P
Siedepunkt: 64 °C (0.7 mmHg)
Dichte: 1.032
Flammpunkt: >110 °C (230 °F)
MDL Number: MFCD00009086
CAS-Nummer: 1638-86-4
EINECS: 216-676-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR