(Diacetoxyiod)benzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Iodobenzene diacetate, (Diacetoxyiodo)benzene
B24531.22
B24531.14
B24531.36
B24531.22EA
217
EUR
B24531.22
B24531.14
B24531.36
(Diacetoxyiod)benzol ≥98%
(Diacetoxyiod)benzol
Purity: 98+%
CAS No.: 3240-34-4
Molecular Formula: C10H11IO4
Formula Weight: 322.10
Melting Point: 159-161°
Beilstein Reference: 1879369
Formel:
C₁₀H₁₁IO₄ Molekulargewicht: 322.1 g/mol Schmelzpunkt: 159…161 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008692 CAS-Nummer: 3240-34-4 EINECS: 221-808-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...