Diphenyliodoniumhexafluorophosphat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A17934.09
A17934.18
A17934.30
A17934.09EA
88.5
EUR
A17934.09
A17934.18
A17934.30
Diphenyliodoniumhexafluorophosphat ≥98%
Diphenyliodonium hexafluorophosphate
Purity: 98%
CAS No.: 58109-40-3
Molecular Formula: C12H10F6IP
Formula Weight: 426.08
Melting Point: 139-144°
UN No.: 2811
MDL No.: MFCD00061398
Formel:
C₁₂H₁₀F₆IP Schmelzpunkt: 139…144 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00061398 CAS-Nummer: 58109-40-3 EINECS: 261-134-5 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...