Dimethyl-3-oxoglutarat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate
Purity: 97%
CAS No.: 1830-54-2
Molecular Formula: C7H10O5
Formula Weight: 174.16
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₇H₁₀O₅ Siedepunkt: 237…241 °C Schmelzpunkt: 16…17 °C Dichte: 1.185 Flammpunkt: >110 °C (230 °F) |
MDL Number:
MFCD00008462 CAS-Nummer: 1830-54-2 EINECS: 217-385-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...