Diethyl-3-oxoglutarat 96%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate
A10933.18
A10933.30
A10933.18EA
69.4
EUR
A10933.18
A10933.30
Diethyl-3-oxoglutarat 96%
Diethyl-3-oxoglutarat
Purity: 96%
CAS No.: 105-50-0
Molecular Formula: C9H14O5
Formula Weight: 202.21
Storage Temperature: 2°C to 8°C
Flash Point: 86°(186°F)
Refractive Index: 1.4400
Boiling Point: 134-135°/10mm
Beilstein Reference: 640146
MDL No.: MFCD00009200
Formel:
C₉H₁₄O₅ Siedepunkt: 134…135 °C (10 mmHg) Dichte: 1.113 Flammpunkt: 86 °C (186 °F) |
MDL Number:
MFCD00009200 CAS-Nummer: 105-50-0 EINECS: 203-302-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...