Natriumsulfat ≥99,0%, Pulver ACS
Lieferant: Thermo Fisher Scientific
Synonyme: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
11560.36 11560.A3 11560.A9
11560.36EA 29.8 EUR
11560.36 11560.A3 11560.A9
Natriumsulfat ≥99,0%, Pulver ACS
Natriumsulfat
In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass. Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
Notes: Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.
In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass
Formel: Na₂SO₄
Molekulargewicht: 142.04 g/mol
Siedepunkt: 1429 °C (1013 hPa)
Schmelzpunkt: 884 °C
Dichte: 2.680
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003504
CAS-Nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 14,8680
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM