Natriumperoxodisulfat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Sodium peroxidisulphate, Sodium persulphate, Disodium peroxodisulphate, Peroxydisulphuric acid disodium salt

54100.A1 54100.A7
54100.A1EA 62.4 EUR
54100.A1 54100.A7
Natriumperoxodisulfat ≥98%
Natriumperoxodisulfat

Purity: 98% CAS No.: 7775-27-1 Molecular Formula: Na2S2O8 Formula Weight: 238.10 Physical Form: Crystalline Density: 2.40 Merck Reference: 14,8656 UN No.: 1505 MDL No.: MFCD00003501

Formel: Na₂S₂O₈
Molekulargewicht: 238.11 g/mol
Schmelzpunkt: 100
Dichte: 2.40
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003501
CAS-Nummer: 7775-27-1
EINECS: 231-892-1
UN-Nummer: 1505
ADR: 5.1 III
Merck Index: 14,8656

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR