Natriumnitrit ≥97% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

14244.14 14244.22 14244.36
14244.14EA 25.8 EUR
14244.14 14244.22 14244.36
Natriumnitrit ≥97% ACS
Sodium nitrite

Purity: 97% min CAS No.: 7632-00-0 Molecular Formula: NaNO2 Formula Weight: 69.00 Physical Form: Granular Density: 2.17 Melting Point: 271° Solubility: Soluble in water Merck Reference: 14,8648 UN No.: 1500 MDL No.: MFCD00011118

Formel: NaNO₂
Siedepunkt: 320 °C (1013 hPa)
Schmelzpunkt: 271 °C
Dichte: 2,17
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011118
CAS-Nummer: 7632-00-0
EINECS: 231-555-9
UN-Nummer: 1500
ADR: 5.1,III
Merck Index: 14,8648

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR