Natriumperiodat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 7790-28-5
Molecular Formula: INaO4
Formula Weight: 213.89
Physical Form: Powder
Density: 3.86
Appearance: White
Odour: Odourless
Melting Point: 300° dec.
Solubility: Soluble in water and acids
Merck Reference: 14,8640
UN No.: 1479
MDL No.: MFCD00003534
Source of periodic acid, as an analytical reagent, and as an oxidizing agent
Formel:
NaIO₄ Siedepunkt: 300 °C (1013 hPa) Schmelzpunkt: 300 °C (Zers.) Dichte: 3,86 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003534 CAS-Nummer: 7790-28-5 EINECS: 232-197-6 UN-Nummer: 1479 ADR: 5.1,II Merck Index: 14,8640 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...