Kaliumiodid 99,0-100,5% Ph. Eur.

Lieferant: VWR Chemicals

Synonyme: Dipotassium diiodide, Kali iodide

26850.230EA 246 EUR
26850.230 26850.296 26850.365 26850.260 26850.460
Kaliumiodid 99,0-100,5% Ph. Eur.
Kaliumiodid
Formel: Kl
Siedepunkt: 1330 °C (1013 hPa)
Schmelzpunkt: 681 °C
Dichte: 3.13 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011405
CAS-Nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Bestellen

Spezifikation Testergebnisse

Assay (calculated on dried substance) 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Alkalinity Passes test
IO3 (Iodate) Passes test
SO4 (Sulphate) Max. 150 ppm
S2O3 (Thiosulphate) Passes test
Heavy metals (as Pb) Max. 10 ppm
Fe (Iron) Max. 20 ppm
Loss on drying (105°C) Max. 1 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR