Kaliumiodid ≥99,995% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Synonyme:
Dipotassium diiodide, Kali iodide
10978.06
10978.14
10978.22
10978.06EA
75.8
EUR
10978.06
10978.14
10978.22
Kaliumiodid ≥99,995% (Metall-Basis)
Kaliumiodid
Purity: 99.995%
CAS No.: 7681-11-0
Molecular Formula: KI
Formula Weight: 166.01
Physical Form: Crystalline
Density: 3.12
Refractive Index: 1.677
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 681°
Boiling Point: 1330°
Merck Reference: 14,7643
MDL No.: MFCD00011405
Formel:
Kl Siedepunkt: 1330 °C Schmelzpunkt: 681 °C Dichte: 3.12 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011405 CAS-Nummer: 7681-11-0 EINECS: 213-659-4 Merck Index: 14,7643 |