Natriumfluorid 98,5-100,5% Ph. Eur.
Lieferant: VWR Chemicals
|
Gefahr
|
27859.293EA
131
EUR
27859.293
27859.464
Natriumfluorid 98,5-100,5% Ph. Eur.
Sodium fluoride
Formel:
NaF Siedepunkt: 1700 °C (1013 hPa) Schmelzpunkt: 993 °C Dichte: 2,8 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003524 CAS-Nummer: 7681-49-4 EINECS: 231-667-8 UN-Nummer: 1690 ADR: 6.1,III Merck Index: 13,08691 |
|
Spezifikation Testergebnisse
Assay (calculated on dried substance) | 98.5 - 100.5 % |
Appearance | Conforms (see CoA/CoS) |
Identification A | Passes test |
Identification B | Passes test |
Identification C | Passes test |
Appearance of solution | Passes test |
Acidity or alkalinity | Passes test |
Cl (Chloride) | Max. 200 ppm |
Fluorosilicates | Passes test |
SO4 (Sulphate) | Max. 200 ppm |
Loss on drying (130°C) | Max. 0.5 % |
Residual solvents | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...