Kaliumchromat ≥99,0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.0% min
CAS No.: 7789-00-6
Molecular Formula: K2CrO4
Formula Weight: 194.19
Physical Form: Crystalline
Density: 2.73
Appearance: Yellow
Odour: Odourless
Melting Point: 975°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,7622
UN No.: 3087
MDL No.: MFCD00011368
In finishing leather, rust proofing of metals, textile mordant, as an analytical lab reagent
Formel:
K₂CrO₄ Siedepunkt: 1000 °C (1013 hPa) Schmelzpunkt: 975 °C Dichte: 2.73 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011368 CAS-Nummer: 7789-00-6 EINECS: 232-140-5 UN-Nummer: 3087 ADR: 5.1,III Merck Index: 14,7622 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...