Lithiumaluminiumhydrid ≥97%, Plätzchen
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Aluminium lithium hydride, Lithium tetrahydridoaluminate
89643.14
89643.36
89643.14EA
71.1
EUR
89643.14
89643.36
Lithiumaluminiumhydrid ≥97%, Plätzchen
Lithiumaluminiumhydrid
Purity: 97%
CAS No.: 16853-85-3
Molecular Formula: LiAlH4
Formula Weight: 37.95
Physical Form: Pellets
Density: 0.917
Appearance: Whote to grey
Odour: Odourless
Melting Point:
Formel:
H₄AlLi Schmelzpunkt: >125 °C (dec.) Dichte: 0.917 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011075 CAS-Nummer: 16853-85-3 EINECS: 240-877-9 UN-Nummer: 1410 ADR: 4.3,I Merck Index: 14,349 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...