4-Ethylcyclohexanon ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
|
L09363.14
L09363.22
L09363.14EA
76.1
EUR
L09363.14
L09363.22
4-Ethylcyclohexanon ≥99%
4-Ethylcyclohexanon
Purity: 99%
CAS No.: 5441-51-0
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 63°(145°F)
Refractive Index: 1.4515
Odour: Characteristic
Boiling Point: 192-194°
Beilstein Reference: 1924964
MDL No.: MFCD00001644
Formel:
C₈H₁₄O Siedepunkt: 192…194 °C Dichte: 0.895 Flammpunkt: 63 °C (145 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00001644 CAS-Nummer: 5441-51-0 EINECS: 226-629-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...