4-Picolylchlorid Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Chloromethyl)pyridine hydrochloride, 4-(Chloromethyl)-pyridinium chloride

M-Clarity™ quality level = MQ100

P43807-100G P43807-10G
SIALP43807-100GEA 139 EUR
SIALP43807-100G SIALP43807-10G
4-Picolylchlorid Hydrochlorid, Sigma-Aldrich®
4-Picolylchlorid Hydrochlorid
Formel: C₆H₇Cl₂N
MDL Number: MFCD00012826
CAS-Nummer: 1822-51-1
EINECS: 217-350-7
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR