Brij® 35, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Polyethylene glycol octadecyl ether, Steareth 20, Polyoxyethylene (20) stearyl ether
P4019-100G
P4019-500G
SIALP4019-100GEA
48.2
EUR
SIALP4019-100G
SIALP4019-500G
Brij® 35, Sigma-Aldrich®
Brij® 35
Formel:
C₁₈H₃₇(OCH₂CH₂)₂₀OH Schmelzpunkt: 44…46 °C (Lit.) |
MDL Number:
MFCD00074177 CAS-Nummer: 9005-00-9 EINECS: 500-017-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...