(2-Chlorethyl)dimethylammoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-(2-Chloroethyl)-N,N-Dimethylammonium Chloride, 2-Dimethylaminoethyl chloride hydrochloride
M-Clarity™ quality level = MQ200
D141208-100G
D141208-500G
D141208-5G
SIALD141208-100GEA
50.1
EUR
SIALD141208-100G
SIALD141208-500G
SIALD141208-5G
(2-Chlorethyl)dimethylammoniumchlorid, Sigma-Aldrich®
2-Chloroethyldimethylammonium chloride
Formel:
C₄H₁₁Cl₂N |
MDL Number:
MFCD00012516 CAS-Nummer: 4584-46-7 EINECS: 224-970-1 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...