1-Ethylpyridiniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethylpyridin-1-ium tetrafluoroborate

713104-50G 713104-5G
SIAL713104-50GEA 497 EUR
SIAL713104-50G SIAL713104-5G
1-Ethylpyridiniumtetrafluoroborat, Sigma-Aldrich®
1-Ethylpyridiniumtetrafluoroborate
Formel: C₇H₁₀BF₄N
MDL Number: MFCD14156009
CAS-Nummer: 350-48-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR