Bis(2-ethylhexyl)adipat, Supelco®

Lieferant: Merck
Achtung

Synonyme: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate

M-Clarity™ quality level = MQ100

525197-1L 525197-5ML
525197-1LEA 175 EUR
525197-1L 525197-5ML
Bis(2-ethylhexyl)adipat, Supelco®
Bis(2-ethylhexyl)adipat
Formel: C₂₂H₄₂O₄
MDL Number: MFCD00009496
CAS-Nummer: 103-23-1
EINECS: 203-090-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3002beed5039b03ce44d28b9cae34d1ed1cbbfe&vid=8f48dc002fadfa6cb454cfde96bcd85eea77e473&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56eabe12246fa30af9b899d01008472f99cd94a0&vid=b9b279336783e33335353565e204ff28cd5cf9a4&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR