Lithiumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt

449504-10G 449504-50G
SIAL449504-10GEA 123 EUR
SIAL449504-10G SIAL449504-50G
Lithiumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
Lithiumbis(trifluormethylsulfonyl)imid
Formel: C₂F₆LiNO₄S₂
MDL Number: MFCD00210017
CAS-Nummer: 90076-65-6
EINECS: 415-300-0
UN-Nummer: 2923
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR