Ytterbium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ytterbium (III) triflate

M-Clarity™ quality level = MQ100

430595-1G 430595-5G
SIAL430595-1GEA 33.5 EUR
SIAL430595-1G SIAL430595-5G
Ytterbium(III)trifluormethansulfonat, Sigma-Aldrich®
Ytterbium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Yb
MDL Number: MFCD06200261
CAS-Nummer: 54761-04-5
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR