Yttrium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Yttrium(III) trifluoromethanesulphonate, Yttrium triflate

M-Clarity™ quality level = MQ100

425745-25G 425745-5G
SIAL425745-25GEA 156 EUR
SIAL425745-25G SIAL425745-5G
Yttrium(III)trifluormethansulfonat, Sigma-Aldrich®
Yttrium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Y
MDL Number: MFCD00209623
CAS-Nummer: 52093-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR