Ethylvinylether stabilisiert, Sigma-Aldrich®
Lieferant: Merck
Stabilisierung:
Stabilised with potassium hydroxide
M-Clarity™ quality level = MQ200
422177-1L
422177-250ML
SIAL422177-1LEA
99.2
EUR
SIAL422177-1L
SIAL422177-250ML
Ethylvinylether stabilisiert, Sigma-Aldrich®
Ethylvinylether
Formel:
C₄H₈O |
MDL Number:
MFCD00009248 CAS-Nummer: 109-92-2 EINECS: 203-718-4 UN-Nummer: 1302 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...