Brij® 56, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

388858-100G 388858-1KG
SIAL388858-100GEA 49.8 EUR
SIAL388858-100G SIAL388858-1KG
Brij® 56, Sigma-Aldrich®
Brij® 56
Formel: C₃₆H₇₄O₁₁
CAS-Nummer: 9004-95-9
EINECS: 500-014-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR