Triphenylphosphindichlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

378755-100G 378755-25G
SIAL378755-100GEA 147 EUR
SIAL378755-100G SIAL378755-25G
Triphenylphosphindichlorid, Sigma-Aldrich®
Triphenylphosphindichlorid
Formel: C₁₈H₁₅Cl₂P
MDL Number: MFCD00010359
CAS-Nummer: 2526-64-9
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR