Kupfer(II)trifluormethansulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Copper(II) trifluoromethanesulphonate, Copper triflate
M-Clarity™ quality level = MQ100
283673-25G
283673-5G
SIAL283673-25GEA
369
EUR
SIAL283673-25G
SIAL283673-5G
Kupfer(II)trifluormethansulfonat, Sigma-Aldrich®
Kupfer(II)trifluormethansulfonat
Formel:
C₂CuF₆O₆S₂ |
MDL Number:
MFCD00077492 CAS-Nummer: 34946-82-2 EINECS: 252-300-8 UN-Nummer: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...