Bis(triphenylphosphin)nickel(II)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dichlorobis(triphenylphosphine)nickel(II), Bis(triphenylphosphine)nickel(II)chloride
M-Clarity™ quality level = MQ100
237116-10G
237116-50G
SIAL237116-10GEA
64.2
EUR
SIAL237116-10G
SIAL237116-50G
Bis(triphenylphosphin)nickel(II)chlorid, Sigma-Aldrich®
Bis(triphenylphosphin)nickel(II)chlorid
Formel:
C₃₆H₃₂Cl₂NiP₂ |
MDL Number:
MFCD00009592 CAS-Nummer: 14264-16-5 EINECS: 238-154-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...