Brij® 35, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
16005-1KG-F
16005-250G-F
SIAL16005-1KG-FEA
121
EUR
SIAL16005-1KG-F
SIAL16005-250G-F
Brij® 35, Sigma-Aldrich®
Brij® 35
Formel:
C₅₈H₁₁₈O₂₄ Molekulargewicht: 1199.56 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00080891 CAS-Nummer: 9002-92-0 EINECS: 500-002-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...