Bis(2-ethylhexyl)adipat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
M-Clarity™ quality level = MQ200
8.01453.0100
8.01453.1000
8.01453.9050
8.01453.9025
8.01453.0100EA
26.3
EUR
8.01453.0100
8.01453.1000
8.01453.9050
8.01453.9025
Bis(2-ethylhexyl)adipat, Sigma-Aldrich®
Bis(2-ethylhexyl)adipat
Formel:
C₂₂H₄₂O₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00009496 CAS-Nummer: 103-23-1 EINECS: 203-090-1 |
Spezifikation Testergebnisse
Solubility |
|
Melting Point | -68 - -66 °C |
Molar Mass | 370.57 g/mol |
Boiling Point | 247 - 252 °C (27 hPa) |
Vapor Pressure |
|
Flash Point | 195 °C |
Refractive Index | 1.447 (20 °C, 589 nm) |
Density | 0.93 g/cm3 (20 °C) |
pH | 7 (H2O, 20 °C) |
Ignition Point | 350 °C DIN 51794 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...