Ethyl-5-brom-2-chlorbenzoat

Lieferant: Thermo Scientific

SB01145.1 SB01145.10 SB01145.50
MAYBSB01145.1EA 0 EUR
MAYBSB01145.1 MAYBSB01145.10 MAYBSB01145.50
Ethyl-5-brom-2-chlorbenzoat
Ethyl-5-brom-2-chlorbenzoat
Formel: C₉H₈BrClO₂
MDL Number: MFCD00013635
CAS-Nummer: 76008-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR