Samarium(III)trifluormethansulfonat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Samarium(III) trifluoromethanesulphonate
L20253.06
L20253.14
L20253.06EA
72.2
EUR
L20253.06
L20253.14
Samarium(III)trifluormethansulfonat 98%
Samarium(III)trifluormethansulfonat
Purity: 98%
CAS No.: 52093-28-4
Molecular Formula: Sm(SO3CF3)3
Formula Weight: 597.56
MDL No.: MFCD00192431
Formel:
C₃F₉O₉S₃Sm Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00192431 CAS-Nummer: 52093-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...