1-Boc-3-hydroxypiperidin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: tert-butyl 3-hydroxypiperidine-1-carboxylate

L17600.03 L17600.06
L17600.03EA 40.9 EUR
L17600.03 L17600.06
1-Boc-3-hydroxypiperidin ≥97%
1-Boc-3-hydroxypiperidin

Purity: 97% CAS No.: 85275-45-2 Molecular Formula: C10H19NO3 Formula Weight: 201.27 Melting Point: 66-70° Beilstein Reference: 4180859

Formel: C₁₀H₁₉NO₃
Schmelzpunkt: 66…70 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02093938
CAS-Nummer: 85275-45-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR