4-Fluor-2-nitrobenzonitril ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H32610.03
H32610.09
H32610.03EA
115
EUR
H32610.03
H32610.09
4-Fluor-2-nitrobenzonitril ≥97%
4-Fluor-2-nitrobenzonitril
Purity: 97%
CAS No.: 80517-21-1
Molecular Formula: C7H3FN2O2
Formula Weight: 166.11
Melting Point: 69-73°
UN No.: 3439
MDL No.: MFCD00277447
Formel:
C₇H₃FN₂O₂ Schmelzpunkt: 69…73 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00277447 CAS-Nummer: 80517-21-1 UN-Nummer: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...