1-Dodecylpyridiniumchlorid Hydrat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23492.14
B23492.22
B23492.14EA
66.6
EUR
B23492.14
B23492.22
1-Dodecylpyridiniumchlorid Hydrat 98%
1-Dodecylpyridiniumchlorid Hydrat
Purity: 98%
CAS No.: 207234-02-4
Molecular Formula: C17H30ClN.xH2O
Formula Weight: 283.89(anhy)
Melting Point: 88-90°
MDL No.: MFCD00150002
Formel:
C₁₇H₃₀ClN·xH₂O Schmelzpunkt: 88…90 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150002 CAS-Nummer: 207234-02-4 EINECS: 203-232-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...