Triethyloxoniumtetrafluoroborat ≥95% stabilisiert
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Stabilisierung:
Stabilised with diethyl ether
A14420.14
A14420.22
A14420.14EA
89.5
EUR
A14420.14
A14420.22
Triethyloxoniumtetrafluoroborat ≥95% stabilisiert
Triethyloxonium tetrafluoroborate
Stabilized with 3-5% diethyl ether
Purity: 95%
CAS No.: 368-39-8
Molecular Formula: C6H15BF4O
Formula Weight: 189.99
Storage Temperature: 2°C to 8°C
Melting Point: 84-89°
Beilstein Reference: 3598090
UN No.: 2925
MDL No.: MFCD00044423
Formel:
C₆H₁₅BF₄O Schmelzpunkt: 84…89 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00044423 CAS-Nummer: 368-39-8 EINECS: 206-705-1 UN-Nummer: 2925 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...