cis-Bis-(triphenylphosphin)-platin(II)-dichlorid ≥99,95% (Metall-Basis) Pt ≥ 24.2%, Premion®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: cis-Bis(triphenylphosphine)palladium (II) chloride, cis-Bis(triphenylphosphine)platinum(II) chloride, Pt(PPh₃)₂Cl₂

41008.06 41008.03
41008.06EA 745 EUR
41008.06 41008.03
cis-Bis-(triphenylphosphin)-platin(II)-dichlorid ≥99,95% (Metall-Basis) Pt ≥ 24.2%, Premion®
cis-Bis-(triphenylphosphin)-platin(II)-dichlorid

Purity: 99.95% CAS No.: 15604-36-1 Molecular Formula: PtCl2[P(C6H5)3]2 Formula Weight: 790.58 Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00010825

Formel: [(C₆H₅)₃P]₂PtCl₂
Molekulargewicht: 790.56 g/mol
Schmelzpunkt: 310 °C (Zers.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010825
CAS-Nummer: 15604-36-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR