Brij® C20
Lieferant: Thermo Fisher Scientific
Synonyme:
Polyoxyethylene(20) cetyl ether
344290050
344295000
344290025
ACRO344290050EA
31.4
EUR
ACRO344290050
ACRO344295000
ACRO344290025
Brij® C20
Brij® C20
Formel:
C₁₆H₃₃(OCH₂CH₂)₂₀OH Schmelzpunkt: 35.4…39.6 °C Flammpunkt: >149 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00080892 CAS-Nummer: 9004-95-9 EINECS: 500-014-1 |
Spezifikation Testergebnisse
Appearance | White Pellets |
Infrared spectrum | Conforms |
Water | ≤1 % |
Additional info | remark: without antioxidant |
Impurity | ≤1 ppm (Free ethyleneoxide) |
Dioxane | ≤10 ppm |
Acid value | ≤1 mg KOH/g |
Hydroxyl value | 45 to 60 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...