4-Hydroxychinaldin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-2-methylquinoline
A14587.06
A14587.14
A14587.22
A14587.06EA
48.2
EUR
A14587.06
A14587.14
A14587.22
4-Hydroxychinaldin ≥98%
4-Hydroxychinaldin
Purity: 98+%
CAS No.: 607-67-0
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 234-236°
Beilstein Reference: 118320
Formel:
C₁₀H₉NO Siedepunkt: 313 °C (1013 hPa) Schmelzpunkt: 234…236 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00006758 CAS-Nummer: 607-67-0 EINECS: 210-140-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...