1,2,4-Trimethylbenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pseudocumene
Purity: 98%
CAS No.: 95-63-6
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 48°(118°F)
Physical Form: Liquid
Density: 0.882
Refractive Index: 1.5040
Odour: Aromatic
Melting Point: -44°
Boiling Point: 169-171°
Solubility: Soluble in alcohol, benzene, ether
Beilstein Reference: 1903005
Merck Reference: 14,7915
UN No.: 3295
MDL No.: MFCD00008527
Formel:
C₉H₁₂ Molekulargewicht: 120.19 g/mol Siedepunkt: 169…171 °C Schmelzpunkt: –44 °C Dichte: 0.882 Flammpunkt: 48 °C (118 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008527 CAS-Nummer: 95-63-6 EINECS: 202-436-9 UN-Nummer: 3295 ADR: 3 III Merck Index: 12,08100 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...