Tetrahexylammoniumhydrogensulfat, Sigma-Aldrich®

Lieferant: Merck

87303-25G 87303-5G
SIAL87303-25GEA 269 EUR
SIAL87303-25G SIAL87303-5G
Tetrahexylammoniumhydrogensulfat, Sigma-Aldrich®
Tetrahexylammonium hydrogen sulphate
Formel: C₂₄H₅₃NO₄S
Molekulargewicht: 451.76 g/mol
MDL Number: MFCD00037675
CAS-Nummer: 32503-34-7
EINECS: 251-069-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR