Boc-Pro-OH, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid

15490-100G 15490-25G 15490-5G
SIAL15490-100GEA 87.6 EUR
SIAL15490-100G SIAL15490-25G SIAL15490-5G
Boc-Pro-OH, Sigma-Aldrich®
Boc-Pro-OH
Formel: C₁₀H₁₇NO₄
MDL Number: MFCD00037324
CAS-Nummer: 15761-39-4
EINECS: 239-848-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR