Boc-D-Pro-OH ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

L08826.03 L08826.06
L08826.03EA 57.9 EUR
L08826.03 L08826.06
Boc-D-Pro-OH ≥98%
Boc-D-Pro-OH

Purity: 98+% CAS No.: 37784-17-1 Molecular Formula: C10H17NO4 Formula Weight: 215.25 Storage Temperature: 2°C to 8°C Melting Point: 133-135° Beilstein Reference: 479316 Optical Rotation: +61° (c=1 in acetic acid) MDL No.: MFCD00063226

Formel: C₁₀H₁₇NO₄
Schmelzpunkt: 133…135 °C
MDL Number: MFCD00063226
CAS-Nummer: 37784-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR