5-Brom-2-thiophencarbaldehyd ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Bromo-2-thiophenecarboxaldehyde

A13381.06 A13381.14 A13381.22
A13381.06EA 44.6 EUR
A13381.06 A13381.14 A13381.22
5-Brom-2-thiophencarbaldehyd ≥97%
5-Brom-2-thiophencarbaldehyd

Purity: 97% CAS No.: 4701-17-1 Molecular Formula: C5H3BrOS Formula Weight: 191.05 Storage Temperature: 2°C to 8°C Flash Point: 98°(208°F) Refractive Index: 1.6390 Boiling Point: 104-106°/10mm Beilstein Reference: 108404 MDL No.: MFCD00005432

Formel: C₅H₃BrOS
Siedepunkt: 104…106 °C (10 mmHg)
Dichte: 1.762
Flammpunkt: 98 °C (208 °F)
MDL Number: MFCD00005432
CAS-Nummer: 4701-17-1
EINECS: 225-176-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR