Boc-Pro-OH 99+%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
275650250
275651000
ACRO275650250EA
60.9
EUR
ACRO275650250
ACRO275651000
Boc-Pro-OH 99+%
Boc-Pro-OH
Formel:
C₁₀H₁₇NO₄ Schmelzpunkt: 133…136 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00037324 CAS-Nummer: 15761-39-4 EINECS: 239-848-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 133 to 136 °C |
Water | ≤0.5 % (K.F.) |
HPLC | ≥99.0 % |
Specific optical rotation | -59.5° to -61.5° (20 °C, 589 nm) (c=2, CH3COOH) |
Titration with NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...