α-Brom-o-xylol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Methylbenzyl bromide, alpha-Bromo-o-xylene

B21717.14 B21717.22
B21717.14EA 95 EUR
B21717.14 B21717.22
α-Brom-o-xylol ≥98%
α-Brom-o-xylol

Purity: 98% CAS No.: 89-92-9 Molecular Formula: C8H9Br Formula Weight: 185.07 Storage Temperature: 2°C to 8°C Flash Point: 82°(179°F) Refractive Index: 1.5750 Melting Point: 18-20° Boiling Point: 216-217° Beilstein Reference: 636333 Merck Reference: 14,10090 UN No.: 1701 MDL No.: MFCD00000174

Formel: C₈H₉Br
Siedepunkt: 216…217 °C
Schmelzpunkt: 18…20 °C
Dichte: 1.381
Flammpunkt: 82 °C (179 °F)
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00000174
CAS-Nummer: 89-92-9
EINECS: 201-951-6
UN-Nummer: 1701
ADR: 6.1,II
Merck Index: 14,10090

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR