α,α'-Dibrom-o-xylol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,2-Bis(bromomethyl)benzene, o-Xylylene bromide, o-Xylylene dibromide, alpha,alpha'-Dibromo-o-xylene, o-Xylene dibromide

A17241.14 A17241.06 A17241.22
A17241.14EA 58.3 EUR
A17241.14 A17241.06 A17241.22
α,α'-Dibrom-o-xylol ≥97%
α,α'-Dibrom-o-xylol

Purity: 97% CAS No.: 91-13-4 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 92-96° Beilstein Reference: 637159 UN No.: 3261 MDL No.: MFCD00000175

Formel: C₈H₈Br₂
Siedepunkt: 140 °C (20 mmHg)
Schmelzpunkt: 92…96 °C
Dichte: 1.960
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00000175
CAS-Nummer: 91-13-4
EINECS: 202-042-7
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR