Ethylbenzol 99.8%, rein

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Phenylethane

118080250 118085000 118080025 118080251 118080010
ACRO118080250EA 21.7 EUR
ACRO118080250 ACRO118085000 ACRO118080025 ACRO118080251 ACRO118080010
Ethylbenzol 99.8%, rein
Ethylbenzol
Formel: C₈H₁₀
Siedepunkt: 136 °C (760 mmHg)
Schmelzpunkt: –95 °C
Dichte: 0.86 g/cm³
Flammpunkt: 22 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011647
CAS-Nummer: 100-41-4
EINECS: 202-849-4
UN-Nummer: 1175
ADR: 3,II
Merck Index: 13,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4970 (20°C, 589 nm)
Water ≤0.02 % (Coulometric)
GC ≥99.75 %
Color scale ≤10 APHA
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Silicon dioxide (SiO2) ≤50 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.01 %
Loss on heating ≤1.0 % (285°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR