4-Ethylanilin 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

118060050 118061000 118065000
ACRO118060050EA 35.5 EUR
ACRO118060050 ACRO118061000 ACRO118065000
4-Ethylanilin 99+%
4-Ethylanilin
Formel: C₈H₁₁N
Siedepunkt: 216 °C (760 mmHg)
Schmelzpunkt: –5 °C
Dichte: 0,97 g/cm³
Flammpunkt: 85 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007921
CAS-Nummer: 589-16-2
EINECS: 209-637-0
UN-Nummer: 2810
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5532 to 1.5552 (20°C, 589 nm)
GC ≥99.0 %
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.015 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR